Learn More
Bellco Glass, Inc. Beaker-Heavy Wall,600mL
SDP
Catalog No. 501945689
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 600mLDimensions: 95 x 110mmCase Quantity: 12
Catalog No. 50-194-5689 Supplier Bellco Glass, Inc. Supplier No. 562000600
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. Heavy wall beakers meet astime-960 type 1 specifications for classification design capacity and markings. Features a easy pour spout, white permanent graduations and a large marking square. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 600mLDimensions: 95 x 110mmCase Quantity: 12