Learn More
Wilmad Labglass BEAKER JACKETED 1000ML
Catalog No. 50806508
Encompass
Beaker, Jacketed. Capacity: 1000mL. Fully jacketed to all temperature control of contents. Similar to a Griffin beaker, except without a pouring spout.
Catalog No. 50-806-508 Supplier Wilmad Labglass Supplier No. LG3300108
Beaker, Jacketed. Capacity: 1000mL. Fully jacketed to all temperature control of contents. Similar to a Griffin beaker, except without a pouring spout.