Learn More
Wilmad Labglass BEAKER JACKETED 100ML
Catalog No. 50803417
Encompass
Beaker, Jacketed. Capacity: 100mL. Fully jacketed to all temperature control of contents. Similar to a Griffin beaker, except without a pouring spout.
Catalog No. 50-803-417 Supplier Wilmad Labglass Supplier No. LG3300100
Beaker, Jacketed. Capacity: 100mL. Fully jacketed to all temperature control of contents. Similar to a Griffin beaker, except without a pouring spout.