Learn More
Wilmad Labglass BEAKER JACKETED600ML
Catalog No. 50806908
Encompass
Beaker, Jacketed. Capacity: 600mL. Fully jacketed to all temperature control of contents. Similar to a Griffin beaker, except without a pouring spout. Approx I.D. x Height mm: 90 x 125
Catalog No. 50-806-908 Supplier Wilmad Labglass Supplier No. LG3300106
Beaker, Jacketed. Capacity: 600mL. Fully jacketed to all temperature control of contents. Similar to a Griffin beaker, except without a pouring spout. Approx I.D. x Height mm: 90 x 125