Learn More
Chemglass Life Sciences Chemglass Beaker, Griffin Low Form, 10 mL (CG-8048-10), Pyrex
SDP
Catalog No. NC1963994
Encompass_Preferred
The Chemglass CG-8048-10 is a 10 mL low-form (Griffin) PYREX beaker intended for general laboratory mixing, heating, and pouring. It features a spout and uniform wall thickness for improved thermal shock resistance and mechanical strength, and is supplied in packs of 12.
  • 10 mL capacity.
  • Made from PYREX borosilicate glass.
  • Low-form (Griffin) design with pouring spout.
  • Not graduated.
  • Outer diameter approximately 25 mm, height approximately 32 mm.
  • Supplied as pack of 12.
Catalog No. NC1963994 Supplier Chemglass Life Sciences Supplier No. CG804810
The Chemglass CG-8048-10 is a 10 mL low-form (Griffin) PYREX beaker intended for general laboratory mixing, heating, and pouring. It features a spout and uniform wall thickness for improved thermal shock resistance and mechanical strength, and is supplied in packs of 12.
  • 10 mL capacity.
  • Made from PYREX borosilicate glass.
  • Low-form (Griffin) design with pouring spout.
  • Not graduated.
  • Outer diameter approximately 25 mm, height approximately 32 mm.
  • Supplied as pack of 12.