Learn More
Chemglass Life Sciences Chemglass Griffin Low Form Beaker, Pyrex, 150 mL, Graduated
SDP
Catalog No. NC1963998
Encompass_Preferred
This 150 mL Griffin low-form beaker is made from borosilicate (Pyrex) glass with uniform wall thickness and a pour spout. It provides thermal shock resistance, clear graduations for approximate volume measurement, and an extra-large marking spot. Suitable for general laboratory mixing, heating, and reagent handling.
  • Low-form Griffin shape for easy access and mixing.
  • 150 mL nominal capacity with graduated scale for approximate measurements.
  • Borosilicate glass construction offers thermal shock resistance and chemical durability.
  • Pour spout for controlled transfer.
  • Extra-large marking spot for sample identification.
  • Available in individual or case pack ordering options.
Catalog No. NC1963998 Supplier Chemglass Life Sciences Supplier No. CG8048150
This 150 mL Griffin low-form beaker is made from borosilicate (Pyrex) glass with uniform wall thickness and a pour spout. It provides thermal shock resistance, clear graduations for approximate volume measurement, and an extra-large marking spot. Suitable for general laboratory mixing, heating, and reagent handling.
  • Low-form Griffin shape for easy access and mixing.
  • 150 mL nominal capacity with graduated scale for approximate measurements.
  • Borosilicate glass construction offers thermal shock resistance and chemical durability.
  • Pour spout for controlled transfer.
  • Extra-large marking spot for sample identification.
  • Available in individual or case pack ordering options.