Learn More
Hach Company Beaker, Griffin, Low Form, Glass, 150 mL, 12/pk
Supplier:  Hach Company 50074
Encompass
Griffin style (low form) beakers are the industry standard for a wide variety of lab applications. Graduations are high quality screen printing. Bull nose lip and molded pour spout minimize spillage in liquid transfer. Autoclavable, non-toxic and biologically inert. Volumes and graduations are approximate.
Catalog No. 50-199-4315