Learn More
Chemglass Life Sciences Chemglass Griffin Low-Form Beaker, 20 mL, Graduated (CG-8048-20)
SDP
Catalog No. NC2943928
Encompass_Preferred
Low-form (Griffin) 20 mL beaker made from borosilicate (Pyrex) glass for routine laboratory mixing, heating, and pouring. The wide mouth and spout allow easy pouring and access for stirring, and the clear graduations provide approximate volume measurement.
  • Borosilicate glass construction for thermal and chemical resistance.
  • Low-form Griffin shape with pouring spout for easy pouring and stirring access.
  • Graduated scale for approximate volume measurement (visible graduations approx. 5-15 mL).
  • Compact dimensions for small-volume tasks (approx. 40 mm height, 32 mm outer diameter).
  • Suitable for mixing, heating, and routine laboratory use.
Catalog No. NC2943928 Supplier Chemglass Life Sciences Supplier No. CG804820
Low-form (Griffin) 20 mL beaker made from borosilicate (Pyrex) glass for routine laboratory mixing, heating, and pouring. The wide mouth and spout allow easy pouring and access for stirring, and the clear graduations provide approximate volume measurement.
  • Borosilicate glass construction for thermal and chemical resistance.
  • Low-form Griffin shape with pouring spout for easy pouring and stirring access.
  • Graduated scale for approximate volume measurement (visible graduations approx. 5-15 mL).
  • Compact dimensions for small-volume tasks (approx. 40 mm height, 32 mm outer diameter).
  • Suitable for mixing, heating, and routine laboratory use.