Learn More
Chemglass Life Sciences Beaker, Low Form, 50mL, Griffin, Graduated
SDP
Catalog No. NC0363062
Encompass_Preferred
Beaker, Low Form, 50mL, Griffin, Graduated,Beaker, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10mL size is not graduated.
Catalog No. NC0363062 Supplier Chemglass Life Sciences Supplier No. CG804850
Add to Cart
Add to Cart
Beaker, Low Form, 50mL, Griffin, Graduated,Beaker, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10mL size is not graduated.