Learn More
Cole-Parmer Cole-Parmer elements Plastic Beaker, Transparent PP, 250 mL, 50/pk
Catalog No. NC1660928 Shop All Cole-Parmer Products
Encompass_Preferred
Featuring pour spouts, molded in graduations, and heat resistance from 5 to 248°F (-15 to 150°C) these polypropylene beakers are excellent for general use in the lab.
Catalog No. NC1660928 Supplier Cole-Parmer Supplier No. 0620012
Add to Cart
Add to Cart
Featuring pour spouts, molded in graduations, and heat resistance from 5 to 248°F (-15 to 150°C) these polypropylene beakers are excellent for general use in the lab.