Learn More
Cole-Parmer Cole-Parmer elements Plastic Beaker, Transparent PP, 600 mL, 25/pk
Catalog No. NC1660929 Shop All Cole-Parmer Products
Encompass_Preferred
Featuring pour spouts, molded in graduations, and heat resistance from 5 to 248F (-15 to 150C) these polypropylene beakers are excellent for general use in the lab.
Catalog No. NC1660929 Supplier Cole-Parmer Supplier No. 0620016
Featuring pour spouts, molded in graduations, and heat resistance from 5 to 248F (-15 to 150C) these polypropylene beakers are excellent for general use in the lab.