Learn More
Chemglass Life Sciences Beaker, Griffin, Polypropylene, 10 Liter, Graduated
SDP
Catalog No. 50872721
Encompass_Preferred
Beaker, Griffin, Polypropylene, 10 Liter, Graduated. Graduated polypropylene beakers with flared tops and a pouring spouts. Steam autoclavable at 121°C.
Catalog No. 50-872-721 Supplier Chemglass Life Sciences Supplier No. CG1108L02
Beaker, Griffin, Polypropylene, 10 Liter, Graduated. Graduated polypropylene beakers with flared tops and a pouring spouts. Steam autoclavable at 121°C.