Learn More
Hach Company Beaker, Polypropylene, Low Form, 400 mL
Supplier:  Hach Company 108048
Encompass
Griffin style (low form) beakers from Nalgene are the industry standard fora wide variety of lab applications. Graduations are high quality screen printing in sizes ≥250mL. Bull nose lip and molded pour spout minimize spillage in liquid transfer. Autoclavable, non-toxic and biologically inert. Volumes and graduations are approximate.
Catalog No. 50-199-4521