Learn More
Hach Company Beaker, Polypropylene, Low Form, 50 mL, 12/pk
Catalog No. 501994524 Shop All Hach Company Products
Encompass
Griffin style (low form) beakers from Nalgene are the industry standard fora wide variety of lab applications. Graduations are high quality screen printing in sizes ≥250mL. Bull nose lip and molded pour spout minimize spillage in liquid transfer. Autoclavable, non-toxic and biologically inert. Volumes and graduations are approximate.
Catalog No. 50-199-4524 Supplier Hach Company Supplier No. 108071
Griffin style (low form) beakers from Nalgene are the industry standard fora wide variety of lab applications. Graduations are high quality screen printing in sizes ≥250mL. Bull nose lip and molded pour spout minimize spillage in liquid transfer. Autoclavable, non-toxic and biologically inert. Volumes and graduations are approximate.