Learn More
Hach Company Polypropylene Low Form Beaker, 600 mL
Supplier:  Hach Company 108052
Encompass
Griffin low form. For general laboratory use. Non-toxic, noncontaminating, and biologically inert. Autoclavable. Combines no drip pouring, durability, and translucency. Approximate volume scales molded in for small sizes. Sizes 250 mL and larger feature silk-screened graduations and double spout for easier pouring. Nalgene.
Catalog No. 50-199-4522