Learn More
Chemglass Life Sciences Beaker, PTFE, 100mL, 68mm OAH X 54mm OD
SDP
Catalog No. 50872723
Encompass_Preferred
Beaker, PTFE, 100mL, 80mm OAH X 52mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.
Catalog No. 50-872-723 Supplier Chemglass Life Sciences Supplier No. CG110902
Beaker, PTFE, 100mL, 80mm OAH X 52mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.