Learn More
Chemglass Life Sciences Beaker, PTFE, 2, 000mL, 210mm OAH X 120mm OD
SDP
Catalog No. 501531007
Encompass_Preferred
Beaker, PTFE, 2,000mL, 215mm OAH X 140mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.
Catalog No. 50-153-1007 Supplier Chemglass Life Sciences Supplier No. CG110906
Beaker, PTFE, 2,000mL, 215mm OAH X 140mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.