Learn More
Ace Glass, Inc. PTFE BEAKER, 400ML
SDP
Catalog No. 50971886
Encompass
400mL flat-bottom beaker made from molded virgin PTFE, with pour spout. Inert, with smooth internal finish.
Catalog No. 50-971-886 Supplier Ace Glass, Inc. Supplier No. 550009
400mL flat-bottom beaker made from molded virgin PTFE, with pour spout. Inert, with smooth internal finish.