Learn More
Ace Glass, Inc. Beaker, PTFE, thermotech bottom, 100mL
SDP
Catalog No. 50971888
Encompass
100mL molded, pure inert PTFE beaker with pour spout. Bottom surface has molded, stabilized PTFE-carbon outer surface for better heat transfer, and to handle higher temperatures up to 270C.
Catalog No. 50-971-888 Supplier Ace Glass, Inc. Supplier No. 550022
Add to Cart
Add to Cart
100mL molded, pure inert PTFE beaker with pour spout. Bottom surface has molded, stabilized PTFE-carbon outer surface for better heat transfer, and to handle higher temperatures up to 270C.