Learn More
Ace Glass, Inc. Beaker, PTFE, thermotech bottom, 250mL
SDP
Catalog No. 50978369
Encompass
250mL molded, pure inert PTFE beaker with pour spout. Bottom surface has molded, stabilized PTFE-carbon outer surface for better heat transfer, and to handle higher temperatures up to 270C.
Catalog No. 50-978-369 Supplier Ace Glass, Inc. Supplier No. 550024
250mL molded, pure inert PTFE beaker with pour spout. Bottom surface has molded, stabilized PTFE-carbon outer surface for better heat transfer, and to handle higher temperatures up to 270C.