Learn More
Cole-Parmer Pyrex 1000-150 Brand 1000 Griffin Low-Form Beaker, 150 mL, 12/pk
Catalog No. NC9370944 Shop All Cole-Parmer Products
Encompass_Preferred
Glass beakers with spouts are manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength. The 250-mL to 4000-mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot.
This item is not returnable. View return policy
Catalog No. NC9370944 Supplier Cole-Parmer Supplier No. 3450206
Glass beakers with spouts are manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength. The 250-mL to 4000-mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot.