Learn More
Cole-Parmer Pyrex 1000-1L Brand 1000 Griffin Low-Form Beaker, 1000 mL, 6/pk
Catalog No. NC9370942 Shop All Cole-Parmer Products
Encompass_Preferred
Glass beakers with spouts are manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength. The 250-mL to 4000-mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot.
This item is not returnable. View return policy
Catalog No. NC9370942 Supplier Cole-Parmer Supplier No. 3450211
Add to Cart
Add to Cart
Glass beakers with spouts are manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength. The 250-mL to 4000-mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot.