Learn More
Chemglass Life Sciences Beaker, Quartz, 1000mL, 140mm OAH X 107mm OD
SDP
Catalog No. 50871938
Encompass_Preferred
Beaker, Quartz, 1000mL, 140mm OAH X 107mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.
Catalog No. 50-871-938 Supplier Chemglass Life Sciences Supplier No. CG111006
Add to Cart
Add to Cart
Beaker, Quartz, 1000mL, 140mm OAH X 107mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.