Learn More
Chemglass Life Sciences Beaker, Quartz, 100mL, 62mm OAH X 51mm OD
SDP
Catalog No. 50871934
Encompass_Preferred
Beaker, Quartz, 100mL, 62mm OAH X 51mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.
Catalog No. 50-871-934 Supplier Chemglass Life Sciences Supplier No. CG111002
Beaker, Quartz, 100mL, 62mm OAH X 51mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.