Learn More
Chemglass Life Sciences Beaker, Quartz, 250mL, 86mm OAH X 67mm OD
SDP
Catalog No. 50871935
Encompass_Preferred
Beaker, Quartz, 250mL, 86mm OAH X 67mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.
Catalog No. 50-871-935 Supplier Chemglass Life Sciences Supplier No. CG111003
Beaker, Quartz, 250mL, 86mm OAH X 67mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.