Learn More
Chemglass Life Sciences Beaker, Quartz, 400mL, 105mm OAH X 80mm OD
SDP
Catalog No. 50871936
Encompass_Preferred
Beaker, Quartz, 400mL, 105mm OAH X 80mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.
Catalog No. 50-871-936 Supplier Chemglass Life Sciences Supplier No. CG111004
Beaker, Quartz, 400mL, 105mm OAH X 80mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.