Learn More
Chemglass Life Sciences Beaker, Quartz, 50mL, 50mm OAH X 45mm OD
SDP
Catalog No. 50871933
Encompass_Preferred
Beaker, Quartz, 50mL, 50mm OAH X 45mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.
Catalog No. 50-871-933 Supplier Chemglass Life Sciences Supplier No. CG111001
Beaker, Quartz, 50mL, 50mm OAH X 45mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.