Learn More
Bellco Glass, Inc. Beaker-Std Wall,1 L
SDP
Catalog No. 501945699
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. MADE IN THE USA Size: 1l Dimensions: 110x150mm
Catalog No. 50-194-5699 Supplier Bellco Glass, Inc. Supplier No. 562101000
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. MADE IN THE USA Size: 1l Dimensions: 110x150mm