Learn More
Bellco Glass, Inc. Beaker-Std Wall,2 L
SDP
Catalog No. 501945700
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. MADE IN THE USA Size: 2l Dimensions: 135x200mm
Catalog No. 50-194-5700 Supplier Bellco Glass, Inc. Supplier No. 562102000
Add to Cart
Add to Cart
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. MADE IN THE USA Size: 2l Dimensions: 135x200mm