Learn More
Bellco Glass, Inc. Beaker-Std Wall,50mL
SDP
Catalog No. 501945693
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. MADE IN THE USA Sizes: 50ml Dimensions: 46x60mm
Catalog No. 50-194-5693 Supplier Bellco Glass, Inc. Supplier No. 562100050
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. MADE IN THE USA Sizes: 50ml Dimensions: 46x60mm