Learn More
Elma™ Beaker Supporting Lid for Elmasonic S50R Ultrasonic Sieve Cleaner
SDP
Catalog No. 08000136
Change view
Click to view available options
For Use With (Application):
Allows Glass Beakers to be Inserted in Bath for Direct Ultrasonic Treatment
Catalog No. For Use With (Application)
08-000-136 Allows Glass Beakers to be Inserted in Bath for Direct Ultrasonic Treatment
1 options
Catalog No. 08-000-136 Supplier Elma™ Supplier No. 104 9787
Add to Cart
Add to Cart
For use with Elmasonic S50R Ultrasonic Sieve Cleaner.
  • Allows glass beakers to be inserted in bath for direct ultrasonic treatment
  • For glass beakers with diameter 90 to 97mm
  • Rubber ring keeps glass beaker in certain position
Specifications
Product Type Ultrasonic Cleaner Beaker Supporting Lid
For Use With (Equipment) Ultrasonic Cleaner, Elmasonic S50R
For Use With (Application) Allows Glass Beakers to be Inserted in Bath for Direct Ultrasonic Treatment
Material Plastic