Learn More
US Plastic Corporation BEAKER -SZ 5L POLYPL-W/HANDLE
SDP
Catalog No. NC9592531
Encompass
Graduated in milliliters, low form beaker with a design for stability. Made of autoclavable polypropylene.
Catalog No. NC9592531 Supplier US Plastic Corporation Supplier No. 77096
Graduated in milliliters, low form beaker with a design for stability. Made of autoclavable polypropylene.