Learn More
Cole-Parmer Borosil BG1060-500 Tall-Form Glass Beaker; 500 mL, 6/Pk
Catalog No. NC0465726 Shop All Cole-Parmer Products
Encompass_Preferred
United Scientific brand beakers are made from Type l borosilicate glass. Beakers feature heavy and uniform wall thickness for enhanced durability. Spout and beaded top is designed for easy pouring.
Catalog No. NC0465726 Supplier Cole-Parmer Supplier No. 6100077
United Scientific brand beakers are made from Type l borosilicate glass. Beakers feature heavy and uniform wall thickness for enhanced durability. Spout and beaded top is designed for easy pouring.