Learn More
Cole-Parmer Pyrex 1060-500 Brand 1060 Berzelius Tall Form Beaker, 500 mL, 6/pk
Catalog No. NC0333835 Shop All Cole-Parmer Products
Encompass_Preferred
Pyrex Brand 1060 Berzelius beakers are tall and feature a spout to facilitate easy pouring. Beakers are graduated to indicate their approximate content. Reference: ASTM E-960.
This item is not returnable. View return policy
Catalog No. NC0333835 Supplier Cole-Parmer Supplier No. 3451305
Pyrex Brand 1060 Berzelius beakers are tall and feature a spout to facilitate easy pouring. Beakers are graduated to indicate their approximate content. Reference: ASTM E-960.