Learn More
Chemglass Life Sciences Beaker, PTFE, 1, 000mL, 155mm OAH X 100mm OD
SDP
Supplier:  Chemglass Life Sciences CG110905
Encompass_Preferred
Beaker, PTFE, 1000mL, 160mm OAH X 105mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.
Catalog No. 50-872-726