Learn More
Chemglass Life Sciences Beaker, PTFE, 250mL, 97mm OAH X 66mm OD
SDP
Catalog No. 50872724
Encompass_Preferred
Beaker, PTFE, 250mL, 100mm OAH X 65mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.
Catalog No. 50-872-724 Supplier Chemglass Life Sciences Supplier No. CG110903
Add to Cart
Add to Cart
Beaker, PTFE, 250mL, 100mm OAH X 65mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.