Learn More
Chemglass Life Sciences Beaker, PTFE, 50mL, 60mm OAH X 43mm OD
SDP
Catalog No. 50872722
Encompass_Preferred
Beaker, PTFE, 50mL, 60mm OAH X 41mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.
Catalog No. 50-872-722 Supplier Chemglass Life Sciences Supplier No. CG110901
Beaker, PTFE, 50mL, 60mm OAH X 41mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.