Learn More
Andwin Scientific BEAKER TRI-CORNER 50ML 100/CS
SDP
Catalog No. NC0438861
Encompass_Preferred
Tri-Pour Graduated Disposable Beakers 50mL. Transparent polypropylene. Three pouring lips for dripless dispensing. Graduated with accuracy of +-5%. May also be used for measuring. Resistant to strong acids and bases organic solvents.
Catalog No. NC0438861 Supplier Andwin Scientific Supplier No. 25384150
Tri-Pour Graduated Disposable Beakers 50mL. Transparent polypropylene. Three pouring lips for dripless dispensing. Graduated with accuracy of +-5%. May also be used for measuring. Resistant to strong acids and bases organic solvents.