Learn More
Andwin Scientific BEAKER TRI-CORNER100ML 100/CS
SDP
Catalog No. NC0438864
Encompass_Preferred
The 100ml tri-pour graduated disposable beaker features 3 pouring lips for dripless dispensing, graduations, polypropylene construction and write-on cardboard caps. It is resistant to most acids, bases, solvents and reagents.
Catalog No. NC0438864 Supplier Andwin Scientific Supplier No. 25384152
The 100ml tri-pour graduated disposable beaker features 3 pouring lips for dripless dispensing, graduations, polypropylene construction and write-on cardboard caps. It is resistant to most acids, bases, solvents and reagents.