Learn More
Research Products International Corp BEAKERplus Combination Beaker and Flask, 1000 ml Capacity, 95 x 222 mm, 2 per Pack
SDP
Catalog No. 501367660
Encompass
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.
Catalog No. 50-136-7660 Supplier Research Products International Corp Supplier No. 164123
Add to Cart
Add to Cart
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.