Learn More
Research Products International Corp BEAKERplus Combination Beaker and Flask, 1200 ml Capacity, 95 x 254 mm, 2 per Pack
SDP
Catalog No. 501367661
Encompass
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.
Catalog No. 50-136-7661 Supplier Research Products International Corp Supplier No. 164124
Add to Cart
Add to Cart
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.