Learn More
Research Products International Corp BEAKERplus Combination Beaker and Flask, 250 ml Capacity, 70 x 107 mm, 6 per Pack
SDP
Catalog No. 501367658
Encompass
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.
Catalog No. 50-136-7658 Supplier Research Products International Corp Supplier No. 164121
Add to Cart
Add to Cart
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.