Learn More
Chemglass Life Sciences Chemglass Pyrex Griffin Low-Form Beaker, 1 L (CG-8048-1L)
SDP
Catalog No. NC2217094
Encompass_Preferred
Chemglass Pyrex 1 L Griffin low-form beaker (CG-8048-1L) is a 1,000 mL borosilicate glass laboratory beaker designed for general mixing, heating, and approximate measuring tasks. It features a pouring spout, uniform wall thickness for improved thermal shock resistance, and double-scale graduations for approximate volume readings.
  • 1 L (1000 mL) capacity.
  • Borosilicate glass construction.
  • Griffin low-form design with pouring spout.
  • Double-scale graduated markings for approximate measurement.
  • Uniform wall thickness for thermal shock resistance.
  • Extra-large marking spot for labeling.
This item is not returnable. View return policy
Catalog No. NC2217094 Supplier Chemglass Life Sciences Supplier No. CG80481L
Chemglass Pyrex 1 L Griffin low-form beaker (CG-8048-1L) is a 1,000 mL borosilicate glass laboratory beaker designed for general mixing, heating, and approximate measuring tasks. It features a pouring spout, uniform wall thickness for improved thermal shock resistance, and double-scale graduations for approximate volume readings.
  • 1 L (1000 mL) capacity.
  • Borosilicate glass construction.
  • Griffin low-form design with pouring spout.
  • Double-scale graduated markings for approximate measurement.
  • Uniform wall thickness for thermal shock resistance.
  • Extra-large marking spot for labeling.