Learn More
Electron Microscopy Sciences Staccup Beakers 120 cc
SDP
Catalog No. 50996316
Encompass_Preferred
Resistant to most chemicals. Beakers have frosted marking area, customary and metric graduations, and anti-drip ridge. Made of heavy gauge polypropylene. Suitable for parafilm cover.
Catalog No. 50-996-316 Supplier Electron Microscopy Sciences Supplier No. 60950
Resistant to most chemicals. Beakers have frosted marking area, customary and metric graduations, and anti-drip ridge. Made of heavy gauge polypropylene. Suitable for parafilm cover.