Learn More
Grainger Nvent Caddy Beam Clamp, Steel, Electrogalvanized, 1/4 in. Rod, 1/2 in. Flange, 50/Pk.
Catalog No. 502785345
Encompass_Preferred
This beam clamp is designed for securing threaded rods to structural steel beams. Constructed from mild steel with an electrogalvanized finish, the clamp features threaded holes on both the back and bottom to provide versatile mounting options. Installation requires the setscrew to be properly tightened and torqued onto the sloped side of the I-beam.
  • Features threaded holes on the back and bottom
  • Constructed from mild steel with electrogalvanized finish
  • Supports 1/4 in. rod size
  • Accommodates up to 0.5 in. max flange thickness
  • Rated for a 100 lbf static load
  • Requires 20 ft·lbf torque for proper installation
  • Compliant with ANSI/UL 2239, cUL, and CSA standards
Catalog No. 50-278-5345 Supplier Grainger Supplier No. 190A14
This beam clamp is designed for securing threaded rods to structural steel beams. Constructed from mild steel with an electrogalvanized finish, the clamp features threaded holes on both the back and bottom to provide versatile mounting options. Installation requires the setscrew to be properly tightened and torqued onto the sloped side of the I-beam.
  • Features threaded holes on the back and bottom
  • Constructed from mild steel with electrogalvanized finish
  • Supports 1/4 in. rod size
  • Accommodates up to 0.5 in. max flange thickness
  • Rated for a 100 lbf static load
  • Requires 20 ft·lbf torque for proper installation
  • Compliant with ANSI/UL 2239, cUL, and CSA standards