Learn More
MSC PRO-SOURCE Beam Clamp, 2,000 lb Capacity, 3-9 in Grip
Catalog No. 5000513090
Encompass
This beam clamp is designed for securely attaching to structural beams, providing a reliable anchor point for various lifting and rigging applications. With a robust design, it offers a substantial capacity for heavy-duty tasks.
  • 2,000 lb capacity for heavy-duty lifting.
  • 3-9 in grip range for versatile attachment to different beam sizes.
  • Provides a secure and temporary anchor point.
  • Suitable for industrial and construction environments.
Catalog No. 50-005-13090 Supplier MSC Supplier No. 74385154
This beam clamp is designed for securely attaching to structural beams, providing a reliable anchor point for various lifting and rigging applications. With a robust design, it offers a substantial capacity for heavy-duty tasks.
  • 2,000 lb capacity for heavy-duty lifting.
  • 3-9 in grip range for versatile attachment to different beam sizes.
  • Provides a secure and temporary anchor point.
  • Suitable for industrial and construction environments.