Learn More
Ace Glass, Inc. BEARING,GLASS,10MM, NO 15
SDP
Catalog No. NC0794551
Encompass_Preferred
Glass bearing design for use with Ace-Thred pressure reactors and our 8075 glass, polished or 8074 bare, stainless steel 10mm stirring shafts. Both the top bushing and bottom coupling are PTFE. Bearing,glass,10mm,#15-#15
Catalog No. NC0794551 Supplier Ace Glass, Inc. Supplier No. 804407
Glass bearing design for use with Ace-Thred pressure reactors and our 8075 glass, polished or 8074 bare, stainless steel 10mm stirring shafts. Both the top bushing and bottom coupling are PTFE. Bearing,glass,10mm,#15-#15