Learn More
Ace Glass, Inc. Bearing, Trubore, glass, 10mm, 29/42
SDP
Catalog No. 50975939
Encompass_Preferred
10mm Trubore glass bearing with 29/42 standard taper joint. Bearing is for use with aluminum packing box in low positive pressure and vacuum applications. Using the graphite-impregnated PTFE packing and ACE Stir-Lube, pressures down to 7 microns have been obtained at 170rpm, using only a mechanical pump. Speeds over 1000rpm are not advisable due to heat generated. Indefinite packing life. Frequent tightening of the packing box is unnecessary once the packing has run in. The high vacuum lubricant trap adapter prevents lubricant from running into stirred material. Use with precision ground, 10mm stirring shaft.
Catalog No. 50-975-939 Supplier Ace Glass, Inc. Supplier No. 805115
10mm Trubore glass bearing with 29/42 standard taper joint. Bearing is for use with aluminum packing box in low positive pressure and vacuum applications. Using the graphite-impregnated PTFE packing and ACE Stir-Lube, pressures down to 7 microns have been obtained at 170rpm, using only a mechanical pump. Speeds over 1000rpm are not advisable due to heat generated. Indefinite packing life. Frequent tightening of the packing box is unnecessary once the packing has run in. The high vacuum lubricant trap adapter prevents lubricant from running into stirred material. Use with precision ground, 10mm stirring shaft.