Learn More
Ace Glass, Inc. Bearing, Trubore, glass, 10mm, 65/40
SDP
Catalog No. 50975908
Encompass_Preferred
10mm glass Trubore bearing with 65/40 interchangeable ground joint for use with ACE precision ground stirring shafts. Recommended top stirring speed when used with Trubore Stir-Lube is 1500 rpm.
Catalog No. 50-975-908 Supplier Ace Glass, Inc. Supplier No. 803855
10mm glass Trubore bearing with 65/40 interchangeable ground joint for use with ACE precision ground stirring shafts. Recommended top stirring speed when used with Trubore Stir-Lube is 1500 rpm.