Learn More
MCR Safety™ BearKat BK1 Safety Glasses with Clear Lens
Catalog No. 19827924 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Catalog No. Lens Coating
19-827-924 Anti-fog, Impact-resistant
1 options
Catalog No. 19-827-924 Supplier MCR Safety™ Supplier No. BK110PF
BearKat™ BK1 series safety glasses with clear lens, MAX6™ anti-fog coating, soft non-slip temple material.
  • Lightweight frames (25 grams) are comfortable and easy to wear
  • Non-Slip soft temple material
  • Passes ANSI Z87.1 anti-fog test
  • Premium MAX6™ Anti-Fog Lens Coating
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
  • Single lens design with integral side shields provides an unobstructed view
  • Strong yet lightweight design
  • Meets ANSI/ISEA Z87.1 2020 standard
  • Meets ANSI Z87.1 2020 standards
  • Meets CAN/CSA Z94.3 standards
Specifications
Lens Coating Anti-fog, Impact-resistant
Lens Tint Clear
Product Type Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection U6
Frame Type Frameless
Autoclavable Not Autoclavable